siRNA Details
Gene Symbol: Epb4.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183428.3 | NP_906273.3 | 14 | 3385 | ||
GenBank mRNA | CB526050.1 | - | 4 | 328 | ||
L00919.1 | - | 15 | 3460 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
