siRNA Details
Gene Symbol: Epb4.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128606.1 | NP_001122078.1 | 7 | 1258 | ||
NM_001128607.1 | NP_001122079.1 | 7 | 1168 | |||
NM_183428.3 | NP_906273.3 | 7 | 2520 | |||
GenBank mRNA | AB370261.1 | - | 6 | 1043 | ||
AK147266.1 | - | 2 | 214 | |||
AK147644.1 | - | 2 | 214 | |||
AK212118.1 | - | 1 | 92 | |||
AK220462.1 | - | 7 | 1165 | |||
BC068138.1 | - | 7 | 1128 | |||
BC079875.1 | - | 7 | 1087 | 6844082 | ||
CN526385.1 | - | 1 | 102 | |||
CN537562.1 | - | 5 | 576 | |||
L00919.1 | - | 8 | 2595 | |||
LK021332.1 | - | 5 | 413 | |||
LK021333.1 | - | 5 | 413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
