siRNA Details
Gene Symbol: Luzp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024452.2 | NP_077772.2 | 4 | 2682 | ||
GenBank mRNA | AK131161.1 | - | 1 | 1200 | ||
AK137669.1 | - | 1 | 307 | |||
BC053451.1 | - | 1 | 722 | |||
BC137784.1 | - | 2 | 2446 | 8860803 | ||
BC137786.1 | - | 2 | 2446 | 8860805 | ||
BC171934.1 | - | 1 | 2575 | 9054218 | ||
L49344.1 | - | 3 | 2543 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
