siRNA Details
Gene Symbol: Gpr157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177366.3 | NP_796340.2 | 3 | 921 | ||
GenBank mRNA | AK089429.1 | - | 3 | 1061 | ||
AK089544.1 | - | 3 | 874 | |||
AK154291.1 | - | 3 | 557 | |||
AK170941.1 | - | 3 | 516 | |||
AK171081.1 | - | 3 | 516 | |||
BC117996.1 | - | 3 | 875 | 40095824 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
