siRNA Details
Gene Symbol: Chd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081376.1 | NP_001074845.1 | 7 | 1238 | ||
NM_029216.2 | NP_083492.2 | 7 | 1238 | |||
GenBank mRNA | AK147563.1 | - | 7 | 1239 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
