siRNA Details
Gene Symbol: Pank4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039866.1 | - | 1 | 42 | ||
AK050894.1 | - | 4 | 484 | |||
BC050089.1 | - | 4 | 482 | 30018732 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
