siRNA Details
Gene Symbol: Ppp2r2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172994.2 | NP_766582.1 | 9 | 1350 | ||
GenBank mRNA | AK032023.1 | - | 9 | 1253 | ||
AK049830.1 | - | 9 | 1236 | |||
AY348866.1 | - | 9 | 1352 | |||
BC059811.1 | - | 9 | 1350 | 6849535 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
