siRNA Details
Gene Symbol: Zfp664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081750.1 | NP_001075219.1 | 4 | 1211 | ||
GenBank mRNA | BC096550.1 | - | 1 | 294 | 30550168 | |
CV563193.1 | - | 1 | 73 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
