siRNA Details
Gene Symbol: Orai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178751.3 | NP_848866.2 | 3 | 1688 | ||
GenBank mRNA | AK036013.1 | - | 3 | 1678 | ||
AK042940.1 | - | 3 | 1689 | |||
AK079619.1 | - | 3 | 1635 | |||
AK087038.1 | - | 1978 | ||||
BC066070.1 | - | 3 | 1635 | 6853893 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
