siRNA Details
Gene Symbol: Clec4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199311.2 | NP_955015.1 | 2 | 251 | ||
GenBank mRNA | AY397675.1 | - | 2 | 186 | ||
BC049354.1 | - | 2 | 241 | 6310008 | ||
CK626014.1 | - | 2 | 251 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
