siRNA Details
Gene Symbol: Zfp384 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252083.1 | NP_001239012.1 | 6 | 769 | ||
NM_175557.5 | NP_780766.3 | 6 | 740 | |||
GenBank mRNA | AK081740.1 | - | 1 | 49 | ||
AK086185.1 | - | 6 | 665 | |||
AK156026.1 | - | 6 | 771 | |||
AK172476.1 | - | 4 | 1159 | |||
BC064013.1 | - | 6 | 604 | 6515698 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
