siRNA Details
Gene Symbol: Ttll13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177765.3 | NP_808433.2 | 15 | 2567 | ||
GenBank mRNA | AK029565.1 | - | 15 | 2566 | ||
AM690757.1 | - | 14 | 2268 | |||
BC145749.1 | - | 14 | 2431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
