siRNA Details
Gene Symbol: Galntl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175032.3 | NP_778197.2 | 3 | 423 | ||
GenBank mRNA | AK080614.1 | - | 3 | 423 | ||
BB641428.1 | - | 3 | 423 | |||
BC111102.1 | - | 4 | 466 | |||
GU220061.1 | - | 3 | 270 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
