siRNA Details
Gene Symbol: Lrrc36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033371.3 | NP_001028543.1 | 8 | 1419 | ||
NM_001170788.1 | NP_001164259.1 | 11 | 1748 | |||
NM_001170789.1 | NP_001164260.1 | 8 | 1425 | |||
GenBank mRNA | AK133042.1 | - | 8 | 1419 | ||
BC036564.1 | - | 6 | 1036 | |||
BC150957.1 | - | 8 | 1411 | 9087868 | ||
BC172120.1 | - | 8 | 1417 | 9054406 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
