siRNA Details
Gene Symbol: Nlrx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163742.1 | NP_001157214.1 | 3, 4 | 296 | ||
NM_001163743.1 | NP_001157215.1 | 3, 4 | 397 | |||
NM_178420.3 | NP_848507.2 | 3, 4 | 385 | |||
GenBank mRNA | AK089260.1 | - | 3, 4 | 389 | ||
AK141296.1 | - | 3, 4 | 366 | |||
AK145780.1 | - | 3, 4 | 406 | |||
AK145988.1 | - | 3, 4 | 386 | |||
AK166689.1 | - | 3, 4 | 223 | |||
AK170088.1 | - | 3, 4 | 340 | |||
BC034204.1 | - | 2, 3 | 179 | |||
BC050054.1 | - | 3, 4 | 292 | 30012053 | ||
BY196718.1 | - | 3, 4 | 296 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
