siRNA Details
Gene Symbol: Klhdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178253.5 | NP_839984.1 | 8 | 757 | ||
GenBank mRNA | AK144621.1 | - | 8 | 697 | ||
AY238938.1 | - | 8 | 757 | |||
BC119567.2 | - | 8 | 735 | 40057793 | ||
BC119568.2 | - | 8 | 741 | 40057794 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
