siRNA Details
Gene Symbol: Phf21b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081166.2 | NP_001074635.2 | 13 | 2770 | ||
GenBank mRNA | AK033659.1 | - | 2 | 1223 | ||
AK046530.1 | - | 11 | 3025 | |||
BC044847.1 | - | 1 | 3622 | |||
BC048859.1 | - | 1 | 3622 | |||
BC059040.1 | - | 1 | 1590 | |||
BC067021.1 | - | 13 | 2414 | |||
BC157925.1 | - | 13 | 2392 | 9088098 | ||
BC158044.1 | - | 13 | 2392 | 9007348 | ||
BC172019.1 | - | 13 | 2392 | 9054305 | ||
BC172145.1 | - | 12 | 2369 | 9054815 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
