siRNA Details
Gene Symbol: Adcy10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173029.3 | NP_766617.2 | 31 | 4583 | ||
GenBank mRNA | AK029849.1 | - | 31 | 4585 | ||
AK133181.1 | - | 30 | 4471 | |||
BC138448.1 | - | 31 | 4464 | 8861468 | ||
BC145283.1 | - | 31 | 4530 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
