siRNA Details
Gene Symbol: Agbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029744.1 | - | 18 | 2641 | ||
AK030138.1 | - | 18 | 2596 | |||
DQ867027.1 | - | 18 | 3565 | |||
DQ867028.1 | - | 18 | 2641 | |||
DQ867029.1 | - | 17 | 3457 | |||
DQ867030.1 | - | 16 | 3403 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
