siRNA Details
Gene Symbol: Agbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178755.3 | NP_848870.2 | 6 | 493 | ||
GenBank mRNA | AK029744.1 | - | 6 | 526 | ||
AK029932.1 | - | 6 | 494 | |||
AK030182.1 | - | 6 | 540 | |||
AK040259.1 | - | 6 | 429 | |||
AK079691.1 | - | 6 | 430 | |||
DQ867026.1 | - | 6 | 526 | |||
DQ867027.1 | - | 6 | 526 | |||
DQ867028.1 | - | 6 | 526 | |||
DQ867029.1 | - | 6 | 526 | |||
DQ867030.1 | - | 5 | 472 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
