siRNA Details
Gene Symbol: Agbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178755.3 | NP_848870.2 | 11, 12 | 1956 | ||
GenBank mRNA | AK029744.1 | - | 11, 12 | 1989 | ||
AK029932.1 | - | 11, 12 | 1957 | |||
AK030138.1 | - | 11, 12 | 1944 | |||
AK030182.1 | - | 11, 12 | 2003 | |||
AK040259.1 | - | 11, 12 | 1892 | |||
AK041286.1 | - | 3, 4 | 365 | |||
AK079691.1 | - | 11, 12 | 1893 | |||
DQ867026.1 | - | 11, 12 | 1989 | |||
DQ867027.1 | - | 11, 12 | 1989 | |||
DQ867028.1 | - | 11, 12 | 1989 | |||
DQ867029.1 | - | 11, 12 | 1989 | |||
DQ867030.1 | - | 10, 11 | 1935 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
