siRNA Details
Gene Symbol: Arntl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289679.1 | NP_001276608.1 | 5, 6 | 487 | ||
NM_001289680.1 | NP_001276609.1 | 5, 6 | 487 | |||
NM_001289681.1 | NP_001276610.1 | 5, 6 | 487 | |||
NM_172309.2 | NP_758513.1 | 5, 6 | 487 | |||
GenBank mRNA | AY005163.1 | - | 5, 6 | 487 | ||
AY014836.1 | - | 5, 6 | 487 | |||
DQ322241.1 | - | 5, 6 | 487 | |||
DQ355143.1 | - | 5, 6 | 487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
