siRNA Details
Gene Symbol: Spib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019866.1 | NP_063919.1 | 6 | 760 | ||
GenBank mRNA | BC141118.1 | - | 6 | 760 | 9055730 | |
U87620.1 | - | 1 | 237 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
