siRNA Details
Gene Symbol: B3gnt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081167.1 | NP_001074636.1 | 2 | 776 | ||
GenBank mRNA | AK140429.1 | - | 1 | 350 | ||
BC039789.1 | - | 2 | 792 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
