siRNA Details
Gene Symbol: Smtnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177776.3 | NP_808444.1 | 2 | 514 | ||
GenBank mRNA | AK051585.1 | - | 2 | 513 | ||
AK085988.1 | - | 2 | 386 | |||
BC037993.1 | - | 2 | 512 | 5064159 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
