siRNA Details
Gene Symbol: Pigs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201406.1 | NP_958808.1 | 3 | 309 | ||
GenBank mRNA | AK033331.1 | - | 3 | 283 | ||
AK039604.1 | - | 3 | 280 | |||
AK043232.1 | - | 3 | 283 | |||
AK045759.1 | - | 3 | 280 | |||
AK053771.1 | - | 3 | 283 | |||
AK142871.1 | - | 3 | 280 | |||
AK151858.1 | - | 3 | 271 | |||
AK152229.1 | - | 3 | 271 | |||
AK153300.1 | - | 3 | 275 | |||
AK158128.1 | - | 3 | 280 | |||
AK166408.1 | - | 3 | 286 | |||
AK169633.1 | - | 3 | 275 | |||
AK216471.1 | - | 1 | 80 | |||
BC058979.1 | - | 3 | 309 | 5716747 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
