siRNA Details
Gene Symbol: Armc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177778.4 | NP_808446.1 | 2 | 474 | ||
GenBank mRNA | AK042345.1 | - | 2 | 310 | ||
AK146249.1 | - | 2 | 436 | |||
BC023126.1 | - | 2 | 474 | 2616092 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
