siRNA Details
Gene Symbol: Marveld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183195.2 | NP_899018.1 | 1 | 1615 | ||
GenBank mRNA | AK081212.1 | - | 1 | 1298 | ||
AK132243.1 | - | 1 | 1606 | |||
BC044794.1 | - | 1 | 41 | |||
BC054384.1 | - | 1 | 1578 | 6475446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
