siRNA Details
Gene Symbol: Tm4sf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160402.1 | NP_001153874.1 | 5 | 873 | ||
GenBank mRNA | BC147727.1 | - | 5 | 873 | 9056676 | |
BC147729.1 | - | 5 | 873 | 9056678 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
