siRNA Details
Gene Symbol: Trpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177781.4 | NP_808449.1 | 18 | 2152 | ||
GenBank mRNA | AK045771.1 | - | 18 | 2153 | ||
AY231177.1 | - | 18 | 2125 | |||
BC120563.1 | - | 18 | 2143 | 40129486 | ||
BC131963.1 | - | 18 | 2143 | 40130240 | ||
KF460461.1 | - | 18 | 2125 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
