siRNA Details
Gene Symbol: Wfdc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012723.2 | NP_001012741.2 | 2, 3 | 202 | ||
GenBank mRNA | AY542488.1 | - | 2, 3 | 202 | ||
AY542489.1 | - | 2, 3 | 202 | |||
BC043944.1 | - | 2, 3 | 196 | |||
BC145970.1 | - | 1, 2 | 149 | 40131273 | ||
BC145972.1 | - | 1, 2 | 149 | 40131275 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
