siRNA Details
Gene Symbol: Gm694 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033374.3 | NP_001028546.1 | 2 | 162 | ||
GenBank mRNA | AK145525.1 | - | 2 | 163 | ||
BC147054.1 | - | 2 | 128 | 9056056 | ||
BC147055.1 | - | 2 | 128 | 9056077 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
