siRNA Details
Gene Symbol: Depdc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025426.2 | NP_001020597.1 | 14 | 1283 | ||
NM_001170567.1 | NP_001164038.1 | 13 | 980 | |||
NM_177786.4 | NP_808454.2 | 13 | 980 | |||
GenBank mRNA | AK052458.1 | - | 13 | 968 | ||
AK078771.1 | - | 13 | 925 | |||
AK129180.1 | - | 12 | 874 | |||
AK136321.1 | - | 13 | 925 | |||
AK166366.1 | - | 14 | 1246 | |||
AK172587.1 | - | 12 | 862 | |||
BC072604.1 | - | 14 | 1283 | 6831032 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
