siRNA Details
Gene Symbol: Depdc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025426.2 | NP_001020597.1 | 29, 30 | 3105 | ||
NM_001170567.1 | NP_001164038.1 | 28, 29 | 2775 | |||
NM_177786.4 | NP_808454.2 | 28, 29 | 2775 | |||
GenBank mRNA | AK052458.1 | - | 28, 29 | 2763 | ||
AK078771.1 | - | 28, 29 | 2720 | |||
AK129180.1 | - | 27, 28 | 2669 | |||
BC072604.1 | - | 29, 30 | 3105 | 6831032 | ||
CX568961.1 | - | 1, 2 | 47 |

More Information
Other Species Targeted

Gene Ontology Categories:
