siRNA Details
Gene Symbol: Armcx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007578.2 | NP_001007579.1 | 3 | 245 | ||
GenBank mRNA | AK148507.1 | - | 3 | 244 | ||
AK160766.1 | - | 3 | 246 | |||
BC027264.1 | - | 3 | 238 | 3492516 | ||
BY777182.1 | - | 3 | 225 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
