siRNA Details
Gene Symbol: Gm5071 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256004.1 | NP_001242933.1 | 1 | 436 |
Gene Symbol: Gm5072 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114678.1 | NP_001108150.1 | 1 | 436 |
