siRNA Details
Gene Symbol: Wfikkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY308804.1 | - | 2 | 778 | ||
BC113144.1 | - | 2 | 778 | 40047070 | ||
JQ080910.1 | - | 2 | 778 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
