siRNA Details
Gene Symbol: Apol7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024848.2 | NP_001020019.2 | 5 | 1127 | ||
GenBank mRNA | BC085284.1 | - | 5 | 1139 | 30750330 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
