siRNA Details
Gene Symbol: Gm715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271548.1 | NP_001258477.1 | 1 | 686 | ||
GenBank mRNA | BU610071.1 | - | 1 | 72 | ||
BY357849.1 | - | 1 | 266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
