siRNA Details
Gene Symbol: Rilp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029938.2 | NP_001025109.1 | 5 | 730 | ||
GenBank mRNA | AK139016.1 | - | 5 | 722 | ||
AK160517.1 | - | 3 | 523 | |||
BC038146.1 | - | 1 | 48 | |||
BC051502.1 | - | 2 | 189 | |||
BC116750.1 | - | 5 | 714 | 40126069 | ||
BC119166.1 | - | 5 | 714 | 8733915 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
