siRNA Details
Gene Symbol: Emilin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291145.1 | NP_001278074.1 | 4 | 1533 | ||
NM_182840.2 | NP_878260.1 | 4 | 1674 | |||
GenBank mRNA | BC054738.1 | - | 4 | 1663 | 6402350 | |
BC057619.1 | - | 4 | 1663 | 6402350 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
