siRNA Details
Gene Symbol: Adam1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172126.2 | NP_742124.2 | 1 | 2089 | ||
GenBank mRNA | AK132801.1 | - | 1 | 1976 | ||
AK132804.1 | - | 1 | 1980 | |||
AK133150.1 | - | 1 | 1990 | |||
AK133286.1 | - | 1 | 1980 | |||
BC052093.1 | - | 1 | 2089 | 5695131 | ||
KF144408.1 | - | 1 | 311 | |||
KF144409.1 | - | 1 | 311 | |||
U22056.1 | - | 1 | 1051 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
