siRNA Details
Gene Symbol: Sbsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083903.1 | NP_001077372.1 | 1 | 389 | ||
NM_172205.3 | NP_757342.2 | 1 | 389 | |||
GenBank mRNA | AK003459.1 | - | 1 | 87 | ||
AK081760.1 | - | 1 | 324 | |||
AY115494.1 | - | 1 | 328 | |||
AY444556.1 | - | 1 | 328 | |||
BC051531.1 | - | 1 | 398 | |||
BC092350.1 | - | 1 | 318 | |||
BC139385.1 | - | 1 | 327 | 8862407 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
