siRNA Details
Gene Symbol: Klk15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174865.1 | NP_777354.1 | 4 | 521 | ||
GenBank mRNA | BC106965.1 | - | 4 | 521 | 40039744 | |
BC106966.2 | - | 4 | 518 | |||
BC107012.1 | - | 4 | 521 | 40047687 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
