siRNA Details
Gene Symbol: Klk14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174866.2 | NP_777355.1 | 3 | 234 | ||
GenBank mRNA | BC044756.2 | - | 2 | 209 | ||
BC128019.1 | - | 3 | 237 | 40136266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
