siRNA Details
Gene Symbol: Gimap9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174960.2 | NP_777620.1 | 2 | 800 | ||
GenBank mRNA | AK041336.1 | - | 2 | 789 | ||
BC038043.1 | - | 2 | 722 | |||
BC096680.1 | - | 2 | 784 | 1345477 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
