siRNA Details
Gene Symbol: Gimap9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174960.2 | NP_777620.1 | 2 | 796 | ||
GenBank mRNA | AK041336.1 | - | 2 | 785 | ||
BC038043.1 | - | 2 | 718 | |||
BC096680.1 | - | 2 | 780 | 1345477 |
