siRNA Details
Gene Symbol: Hist1h4i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011560.1 | - | 1 | 410 | ||
BC052219.1 | - | 1 | 382 | |||
BC092144.1 | - | 1 | 399 | 30612926 | ||
BC125600.1 | - | 1 | 393 | 40130115 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
