siRNA Details
Gene Symbol: Hist2h2bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175666.2 | NP_783597.2 | 1 | 215 | ||
GenBank mRNA | BC019122.1 | - | 202 | 5003093 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
