siRNA Details
Gene Symbol: Nol4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161483.1 | NP_001154955.1 | 3, 4 | 361 | ||
NM_199024.2 | NP_950189.2 | 3, 4 | 2131 | |||
GenBank mRNA | AK031008.1 | - | 3, 4 | 2131 | ||
BC056377.1 | - | 3, 4 | 354 | 6850243 |
